1-Butanesulphonic acid sodium salt ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A12654.06 A12654.14 A12654.22
A12654.06EA 47.6 EUR
A12654.06 A12654.14 A12654.22
1-Butanesulphonic acid sodium salt ≥99%
1-Butanesulphonic acid sodium salt

Purity: 99% CAS No.: 2386-54-1 Molecular Formula: CH3(CH2)3SO3Na Formula Weight: 160.17 Melting Point: 300° Beilstein Reference: 3716662

Formule: C₄H₉NaO₃S
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007540
CAS nummer: 2386-54-1
EINECS: 219-201-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR