1-Phenyl-2-tetrazoline-5-thione ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A12656.14 A12656.22 A12656.36
A12656.14EA 41.3 EUR
A12656.14 A12656.22 A12656.36
1-Phenyl-2-tetrazoline-5-thione ≥99%
1-Phenyl-2-tetrazoline-5-thione

Purity: 99% CAS No.: 86-93-1 Molecular Formula: C7H6N4S Formula Weight: 178.22 Flash Point: 138°(280°F) Odour: Odourless Beilstein Reference: 139068 UN No.: 1325 MDL No.: MFCD00003129

Formule: C₇H₆N₄S
Vlampunt: 138 °C (280 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003129
CAS nummer: 86-93-1
EINECS: 201-710-5
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR