2-Bromo-3-hydroxypyridine ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A12687.06 A12687.14 A12687.22
A12687.06EA 45.8 EUR
A12687.06 A12687.14 A12687.22
2-Bromo-3-hydroxypyridine ≥99%
2-Bromo-3-hydroxypyridine

Purity: 99% CAS No.: 6602-32-0 Molecular Formula: C5H4BrNO Formula Weight: 174.00 Melting Point: 182-186° Beilstein Reference: 109829

Formule: C₅H₄BrNO
Kookpunt: 232 °C (1013 hPa)
Smeltpunt: 182…186 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006220
CAS nummer: 6602-32-0
EINECS: 229-547-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR