2-Methylbenzyl alcohol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A12696.09 A12696.18 A12696.30
A12696.09EA 38.1 EUR
A12696.09 A12696.18 A12696.30
2-Methylbenzyl alcohol ≥98%
2-Methylbenzyl alcohol

Purity: 98% CAS No.: 89-95-2 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 104°(219°F) Refractive Index: 1.5408 Melting Point: 33-37° Boiling Point: 219° Beilstein Reference: 1929783 MDL No.: MFCD00004622

Formule: C₈H₁₀O
Kookpunt: 219 °C
Smeltpunt: 33…37 °C
Dichtheid: 1,08 g/cm³ (20 °C)
Vlampunt: 104 °C (219 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004622
CAS nummer: 89-95-2
EINECS: 201-954-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR