1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

A12747.06 A12747.14 A12747.22 A12747.99
A12747.06EA 35.5 EUR
A12747.06 A12747.14 A12747.22 A12747.99
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
1,1,1,3,3,3-Hexafluoro-2-propanol

Purity: 99+% CAS No.: 920-66-1 Molecular Formula: C3H2F6O Formula Weight: 168.04 Refractive Index: 1.2750 Melting Point: -4° Boiling Point: 59-60° Beilstein Reference: 1841007 UN No.: 1760 MDL No.: MFCD00011651

Formule: C₃H₂F₆O
Kookpunt: 59…60 °C
Smeltpunt: –4 °C
Dichtheid: 1,619
Vlampunt: >100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011651
CAS nummer: 920-66-1
EINECS: 213-059-4
UN: 1760
ADR: 8,II
Merck Index: 13,09598

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR