Broomethaan ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A12804.0E A12804.22 A12804.36 A12804.PG
A12804.0EEA 104 EUR
A12804.0E A12804.22 A12804.36 A12804.PG
Broomethaan ≥98%
Broomethaan

Purity: 98% CAS No.: 74-96-4 Molecular Formula: C2H5Br Formula Weight: 108.97 Flash Point: -23°(-9°F) Physical Form: Clear liquid Density: 1.461 Refractive Index: 1.4235 Sensitivity: Light Sensitive Appearance: Colourless Odour: Ether-like Melting Point: -119° Boiling Point: 37-39° Beilstein Reference: 1209224 Merck Reference: 14,3771 UN No.: 1891 MDL No.: MFCD00000232

Formule: BrH₂CCH₃
Kookpunt: 37…39 °C
Smeltpunt: –119 °C
Dichtheid: 1,461
Vlampunt: –23 °C (–9 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000232
CAS nummer: 74-96-4
EINECS: 200-825-8
UN: 1891
ADR: 6.1,II
Merck Index: 14,3771

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR