Potassium sorbate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

A12844.0B A12844.0I A12844.30
A12844.0BEA 91.3 EUR
A12844.0B A12844.0I A12844.30
Potassium sorbate ≥99%
Potassium sorbate

Purity: 99% CAS No.: 24634-61-5 Molecular Formula: C6H7KO2 Formula Weight: 150.22 pH: 8,3 Melting Point: 300° MDL No.: MFCD00016546

Formule: CH₃CH=CHCH=CHCOOK
Smeltpunt: >300 °C
Dichtheid: 1,361
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016546
CAS nummer: 24634-61-5
EINECS: 246-376-1
Merck Index: 14,7671
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR