(1-Propyl)triphenylphosphonium bromide ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A12968.18 A12968.30
A12968.18EA 44.3 EUR
A12968.18 A12968.30
(1-Propyl)triphenylphosphonium bromide ≥99%
(1-Propyl)triphenylphosphonium bromide

Purity: 99% CAS No.: 6228-47-3 Molecular Formula: CH3(CH2)2P(C6H5)3Br Formula Weight: 385.29 Melting Point: 236-240° Beilstein Reference: 3599726

Formule: C₂₁H₂₂BrP
Smeltpunt: 236…240 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011843
CAS nummer: 6228-47-3
EINECS: 228-330-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR