Benzyl carbazate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A13005.06 A13005.14 A13005.22
A13005.06EA 52.8 EUR
A13005.06 A13005.14 A13005.22
Benzyl carbazate ≥98%
Benzyl carbazate

Purity: 98+% CAS No.: 5331-43-1 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Melting Point: 66-70° Beilstein Reference: 1952982

Formule: C₈H₁₀N₂O₂
Smeltpunt: 66…70 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041890
CAS nummer: 5331-43-1
EINECS: 226-230-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR