2-(1-Piperidinyl)aniline ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-(1-Piperidino)aniline

A13073.03 A13073.06 A13073.14
A13073.03EA 22.9 EUR
A13073.03 A13073.06 A13073.14
2-(1-Piperidinyl)aniline ≥98%
2-(1-Piperidinyl)aniline

Purity: 98% CAS No.: 39643-31-7 Molecular Formula: C11H16N2 Formula Weight: 176.26 Melting Point: 44-47° Boiling Point: 96-98°/1mm UN No.: 2811 MDL No.: MFCD00047467

Formule: C₁₁H₁₆N₂
MW: 176.26 g/mol
Kookpunt: 96…98 °C (1 mmHg)
Smeltpunt: 44…47 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00047467
CAS nummer: 39643-31-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR