Methyl-1-methyl-1H-pyrrole-2-carboxylate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methyl 1-methylpyrrole-2-carboxylate

A13095.06 A13095.14
A13095.06EA 91.4 EUR
A13095.06 A13095.14
Methyl-1-methyl-1H-pyrrole-2-carboxylate 99%
Methyl-1-methyl-1H-pyrrole-2-carboxylate

Purity: 99% CAS No.: 37619-24-2 Molecular Formula: C7H9NO2 Formula Weight: 139.16 Refractive Index: 1.5220 Boiling Point: 96-98°/15mm Beilstein Reference: 116477 MDL No.: MFCD00052747

Formule: C₇H₉NO₂
MW: 139.16 g/mol
Kookpunt: 96…98 °C (15 mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00052747
CAS nummer: 37619-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR