Hexamethyldisilane 98

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A13155.14 A13155.22
A13155.14EA 89.9 EUR
A13155.14 A13155.22
Hexamethyldisilane 98
Hexamethyldisilane

Purity: 98+% CAS No.: 1450-14-2 Molecular Formula: (CH3)3SiSi(CH3)3 Formula Weight: 146.38 Flash Point: -1°(30°F) Refractive Index: 1.4220 Odour: Odourless Melting Point: 12-13° Boiling Point: 112-113° Beilstein Reference: 1633463 UN No.: 1993 MDL No.: MFCD00008258

Formule: C₆H₁₈Si₂
MW: 146.38 g/mol
Kookpunt: 112…113 °C
Smeltpunt: 12…13 °C
Dichtheid: 0.721
Vlampunt: –1 °C (30 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008258
CAS nummer: 1450-14-2
EINECS: 215-911-0
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR