Phthalaldehyde ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

A13299.18 A13299.09 A13299.30
A13299.18EA 231 EUR
A13299.18 A13299.09 A13299.30
Phthalaldehyde ≥98%
Phthalaldehyde

Purity: 98% CAS No.: 643-79-8 Molecular Formula: C8H6O2 Formula Weight: 134.14 Flash Point: 132°(269°F) Melting Point: 53-57° Beilstein Reference: 878317 UN No.: 2923 MDL No.: MFCD00003335

Formule: C₈H₆O₂
Kookpunt: 266 °C (1013 hPa)
Smeltpunt: 53…57 °C
Dichtheid: 1,13 g/cm³ (20 °C)
Vlampunt: 132 °C (269 °F)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003335
CAS nummer: 643-79-8
EINECS: 211-402-2
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR