L(+)-Arginine Ph. Eur., USP, pure

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A1345.0500 A1345.1000
A1345.0500EA 116 EUR
A1345.0500 A1345.1000
L(+)-Arginine Ph. Eur., USP, pure
L(+)-Arginine
Formule: C₆H₁₄N₄O₂
Kookpunt: 368 °C (1013 hPa)
Smeltpunt: 238 °C
Dichtheid: 1,3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002635
CAS nummer: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR