2-Chloorbenzaldehyde ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A13488.30 A13488.09 A13488.0B
A13488.30EA 27.4 EUR
A13488.30 A13488.09 A13488.0B
2-Chloorbenzaldehyde ≥97%
2-Chloorbenzaldehyde

Purity: 97% CAS No.: 89-98-5 Molecular Formula: C7H5ClO Formula Weight: 140.57 Flash Point: 87°(188°F) Refractive Index: 1.5675 Odour: Pungent pH: 2.9 Melting Point: 10-12° Boiling Point: 212° Beilstein Reference: 385877 UN No.: 3265 MDL No.: MFCD00003304

Formule: C₇H₅ClO
Kookpunt: 212 °C
Smeltpunt: 10…12 °C
Dichtheid: 1,249
Vlampunt: 87 °C (188 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003304
CAS nummer: 89-98-5
EINECS: 201-956-3
UN: 3265
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR