Succinimide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A13503.0B A13503.0I A13503.30
A13503.0BEA 126 EUR
A13503.0B A13503.0I A13503.30
Succinimide ≥98%
Succinimide

Purity: 98+% CAS No.: 123-56-8 Molecular Formula: C4H5NO2 Formula Weight: 99.09 Flash Point: 201°(393°F) Odour: Odourless pH: 4-6 Melting Point: 121-126° Boiling Point: 287-288° Beilstein Reference: 108440 Merck Reference: 14,8871 MDL No.: MFCD00005495

Formule: C₄H₅NO₂
Kookpunt: 287…288 °C
Smeltpunt: 121…126 °C
Dichtheid: 1,410
Vlampunt: 201 °C (393 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005495
CAS nummer: 123-56-8
EINECS: 204-635-6
Merck Index: 14,8871

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR