L(+)-Aspartic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D

A13520.0B A13520.0I A13520.30
A13520.0BEA 94.5 EUR
A13520.0B A13520.0I A13520.30
L(+)-Aspartic acid ≥98%
L(+)-Aspartic acid

Purity: 98+% CAS No.: 56-84-8 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Odour: Odourless Melting Point: 300° dec. Beilstein Reference: 1723530 Merck Reference: 14,840 Optical Rotation: +25° (c=2 in 5N HCl) MDL No.: MFCD00002616

Formule: C₄H₇NO₄
Kookpunt: 311 °C (1013 hPa)
Dichtheid: 1,66
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002616
CAS nummer: 56-84-8
EINECS: 200-291-6
Merck Index: 14,840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR