Guanidiniumchloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

A13543.0I A13543.0B A13543.30
A13543.0IEA 284 EUR
A13543.0I A13543.0B A13543.30
Guanidiniumchloride ≥98%
Guanidiniumchloride

Purity: 98% CAS No.: 50-01-1 Molecular Formula: CH5N3.HCl Formula Weight: 95.53 Melting Point: 178-185° Beilstein Reference: 3591990 Merck Reference: 14,4562 MDL No.: MFCD00013026

Formule: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Smeltpunt: 178…185 °C
Dichtheid: 1.344
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013026
CAS nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 14,4562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR