2-Phenylpropionitrile 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A13636.09 A13636.14
A13636.09EA 53.7 EUR
A13636.09 A13636.14
2-Phenylpropionitrile 96%
2-Phenylpropionitrile

Purity: 97% CAS No.: 1823-91-2 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 93°(199°F) Refractive Index: 1.5106 Boiling Point: 230-232° Beilstein Reference: 774576 UN No.: UN3276 MDL No.: MFCD00001871

Formule: C₉H₉N
Kookpunt: 230…232 °C
Dichtheid: 0,985
Vlampunt: 93 °C (199 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001871
CAS nummer: 1823-91-2
EINECS: 217-354-9
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR