Diethyl (iodomethyl)phosphonate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A13697.06 A13697.14 A13697.22
A13697.06EA 76.8 EUR
A13697.06 A13697.14 A13697.22
Diethyl (iodomethyl)phosphonate ≥98%
Diethyl (iodomethyl)phosphonate

Purity: 98+% CAS No.: 10419-77-9 Molecular Formula: C5H12IO3P Formula Weight: 278.03 Flash Point: 110°(230°F) Refractive Index: 1.4995 Boiling Point: 112-114°/1mm Beilstein Reference: 1812052 MDL No.: MFCD00041386

Formule: C₅H₁₂IO₃P
MW: 278.03 g/mol
Kookpunt: 112…114 °C (1 mmHg)
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041386
CAS nummer: 10419-77-9
EINECS: 233-895-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR