L(-)-Malic acid ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid

A13702.22 A13702.30 A13702.36
A13702.22EA 58.2 EUR
A13702.22 A13702.30 A13702.36
L(-)-Malic acid ≥97%
L(-)-Malic acid
Formule: C₄H₆O₅
Kookpunt: 322 °C (1013 hPa)
Smeltpunt: 100…106 °C
Dichtheid: 1,60
Vlampunt: 220 °C (428 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064213
CAS nummer: 97-67-6
EINECS: 202-601-5
Merck Index: 14,5707

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR