Ethyl-3,4,5-trihydroxybenzoate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A13733.09 A13733.18
A13733.09EA 40.7 EUR
A13733.09 A13733.18
Ethyl-3,4,5-trihydroxybenzoate ≥98%
Ethyl-3,4,5-trihydroxybenzoate

Purity: 98% CAS No.: 831-61-8 Molecular Formula: C9H10O5 Formula Weight: 198.18 Melting Point: 148-152° Beilstein Reference: 2116014

Formule: C₉H₁₀O₅
Smeltpunt: 148…152 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016430
CAS nummer: 831-61-8
EINECS: 212-608-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR