DL-Tyrosine ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A13740.09 A13740.18 A13740.30
A13740.09EA 35.6 EUR
A13740.09 A13740.18 A13740.30
DL-Tyrosine ≥98%
DL-Tyrosine

Purity: 98% CAS No.: 556-03-6 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: ca 325° dec. Beilstein Reference: 515881 Merck Reference: 14,9839 MDL No.: MFCD00063074

Formule: C₉H₁₁NO₃
Kookpunt: 378 °C (1013 hPa)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063074
CAS nummer: 556-03-6
EINECS: 209-113-1
Merck Index: 14,9839

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR