Ethyl-2-furoate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A13909.14 A13909.22 A13909.36
A13909.14EA 22.3 EUR
A13909.14 A13909.22 A13909.36
Ethyl-2-furoate ≥99%
Ethyl-2-furoate

Purity: 99% CAS No.: 614-99-3 Molecular Formula: C7H8O3 Formula Weight: 140.14 Storage Temperature: 2°C to 8°C Flash Point: 70°(158°F) Melting Point: 31-36° Boiling Point: 195-196° Beilstein Reference: 2653 Merck Reference: 14,4307 MDL No.: MFCD00003237

Formule: C₇H₈O₃
Kookpunt: 195…196 °C
Smeltpunt: 31…36 °C
Dichtheid: 1,120
Vlampunt: 70 °C (158 °F)
MDL Nummer: MFCD00003237
CAS nummer: 614-99-3
EINECS: 210-404-0
Merck Index: 14,4307

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR