Ammoniumcarbonaat (∼30% NH₃ )

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Salt of hartshorn

A13955.0B A13955.0I
A13955.0BEA 35.9 EUR
A13955.0B A13955.0I
Ammoniumcarbonaat (∼30% NH₃ )
Ammonium carbonate

CAS No.: 506-87-6 Molecular Formula: CH8N2O3 Formula Weight: 96.09 Odour: Ammonia-like Melting Point: 60° subl. Beilstein Reference: 3627235 Merck Reference: 14,508 MDL No.: MFCD00010890

Formule: (NH₄)₂CO₃
MW: 96.09 g/mol
Smeltpunt: 60 °C (sublimatie)
Dichtheid: 1.5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010890
CAS nummer: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR