α,α'-Dichloro-p-xylene ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A13976.14 A13976.22
A13976.14EA 27.8 EUR
A13976.14 A13976.22
α,α'-Dichloro-p-xylene ≥98%
α,α'-Dichloro-p-xylene

Purity: 98% CAS No.: 623-25-6 Molecular Formula: C8H8Cl2 Formula Weight: 175.06 Melting Point: 98-102° Boiling Point: 254° Solubility: Insoluble in water Beilstein Reference: 774706 UN No.: 2928 MDL No.: MFCD00000920

Formule: C₈H₈Cl₂
Kookpunt: 254 °C
Smeltpunt: 98…102 °C
Dichtheid: 1,417
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000920
CAS nummer: 623-25-6
EINECS: 210-782-7
UN: 2928
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR