4,4'-Dibromobiphenyl (PBB No. 15) ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A14139.18 A14139.30
A14139.18EA 41.4 EUR
A14139.18 A14139.30
4,4'-Dibromobiphenyl (PBB No. 15) ≥98%
4,4'-Dibromobiphenyl (PBB No. 15)

Purity: 98+% CAS No.: 92-86-4 Molecular Formula: C12H8Br2 Formula Weight: 312.02 Melting Point: 164-169° Boiling Point: 355-360° UN No.: 3152 MDL No.: MFCD00000101

Formule: C₁₂H₈Br₂
Kookpunt: 355…360 °C
Smeltpunt: 164…169 °C
Dichtheid: 1,867 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000101
CAS nummer: 92-86-4
EINECS: 202-198-6
UN: 3152
ADR: 9,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR