Cyclopentanecarbonyl chloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A14142.06 A14142.14 A14142.22
A14142.06EA 67.8 EUR
A14142.06 A14142.14 A14142.22
Cyclopentanecarbonyl chloride ≥98%
Cyclopentanecarbonyl chloride

Purity: 98% CAS No.: 4524-93-0 Molecular Formula: C6H9ClO Formula Weight: 132.59 Flash Point: 60°(140°F) Refractive Index: 1.4640 Odour: Acrid Boiling Point: 160-162° Beilstein Reference: 1237147 UN No.: 2920 MDL No.: MFCD00001370

Formule: C₆H₉ClO
Kookpunt: 160…162 °C
Dichtheid: 1,091
Vlampunt: 60 °C (140 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001370
CAS nummer: 4524-93-0
EINECS: 224-856-1
UN: 2920
ADR: 8,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR