α,α,α-Trifluoro-p-toluoyl chloride ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A14176.06 A14176.14
A14176.06EA 61.4 EUR
A14176.06 A14176.14
α,α,α-Trifluoro-p-toluoyl chloride ≥97%
α,α,α-Trifluoro-p-toluoyl chloride

Purity: 98% CAS No.: 329-15-7 Molecular Formula: C8H4ClF3O Formula Weight: 208.57 Flash Point: 78°(172°F) Refractive Index: 1.4770 Odour: Pungent Melting Point: -3° Boiling Point: 188-190° Beilstein Reference: 391282 UN No.: 3265 MDL No.: MFCD00000694

Formule: C₈H₄ClF₃O
Kookpunt: 188…190 °C
Smeltpunt: –3 °C
Dichtheid: 1,404
Vlampunt: 78 °C (172 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000694
CAS nummer: 329-15-7
EINECS: 206-342-9
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR