2-Chloro-5-nitrobenzyl alcohol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A14205.22 A14205.14 A14205.06
A14205.22EA 278 EUR
A14205.22 A14205.14 A14205.06
2-Chloro-5-nitrobenzyl alcohol ≥98%
2-Chloro-5-nitrobenzyl alcohol

Purity: 98% CAS No.: 80866-80-4 Molecular Formula: C7H6ClNO3 Formula Weight: 187.58 Melting Point: 75-79° Beilstein Reference: 3248759

Formule: C₇H₆ClNO₃
Smeltpunt: 75…79 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007297
CAS nummer: 80866-80-4
EINECS: 279-584-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR