(p-Toluenesulphonyl)methyl isocyanide 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A14312.06 A14312.14 A14312.22
A14312.06EA 35.6 EUR
A14312.06 A14312.14 A14312.22
(p-Toluenesulphonyl)methyl isocyanide 97%
(p-Toluenesulphonyl)methyl isocyanide

Purity: 97+% CAS No.: 36635-61-7 Molecular Formula: C9H9NO2S Formula Weight: 195.24 Melting Point: 109-114° Beilstein Reference: 3592382 UN No.: 2811 MDL No.: MFCD00000005

Formule: C₉H₉NO₂S
Smeltpunt: 109…114 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000005
CAS nummer: 36635-61-7
EINECS: 253-140-1
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR