Potassium nitrate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Nitrate of Potash

A14527.36 A14527.0C A14527.0E
A14527.36EA 24.3 EUR
A14527.36 A14527.0C A14527.0E
Potassium nitrate ≥99%
Potassium nitrate

Purity: 99% CAS No.: 7757-79-1 Molecular Formula: KNO3 Formula Weight: 101.11 Odour: Odourless Melting Point: 334° Boiling Point: 400° dec. Merck Reference: 14,7648 UN No.: 1486 MDL No.: MFCD00011409

Formule: KNO₃
Kookpunt: 400 °C (ontbinding)
Smeltpunt: 334 °C
Dichtheid: 2,109
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011409
CAS nummer: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
Bereik: 01-2119488224-35
Merck Index: 14,7648

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR