Nicotinonitrile ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A14850.0E A14850.22 A14850.36
A14850.0EEA 430 EUR
A14850.0E A14850.22 A14850.36
Nicotinonitrile ≥98%
Nicotinonitrile

Purity: 98% CAS No.: 100-54-9 Molecular Formula: C6H4N2 Formula Weight: 104.11 Flash Point: 84°(183°F) Odour: Pungent Boiling Point: 206-208° Beilstein Reference: 107711 MDL No.: MFCD00006372

Formule: C₆H₄N₂
Kookpunt: 206…208 °C
Smeltpunt: 48…51 °C
Dichtheid: 1,16
Vlampunt: 84 °C (183 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00087130
CAS nummer: 100-54-9
EINECS: 202-863-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR