Adenine ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A14906.14 A14906.18 A14906.30 A14906.36 A14906.A7
A14906.14EA 57.1 EUR
A14906.14 A14906.18 A14906.30 A14906.36 A14906.A7
Adenine ≥99%
Adenine

Purity: 99% CAS No.: 73-24-5 Molecular Formula: C5H5N5 Formula Weight: 135.13 Odour: Odourless Melting Point: 360° dec. Beilstein Reference: 5777 Merck Reference: 14,152 UN No.: 2811 MDL No.: MFCD00041790

Formule: C₅H₅N₅
Dichtheid: 0,99741 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041790
CAS nummer: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR