Allylalcohol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A15026.AP A15026.AE A15026.AU
A15026.APEA 60.1 EUR
A15026.AP A15026.AE A15026.AU
Allylalcohol ≥98%
Allylalcohol

Purity: 98+% CAS No.: 107-18-6 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: 22°(71°F) Refractive Index: 1.4130 Odour: Pungent Melting Point: -129° Boiling Point: 95-97° Beilstein Reference: 605307 Merck Reference: 14,286 UN No.: 1098 MDL No.: MFCD00002920

Formule: C₃H₆O
Kookpunt: 95…97 °C
Smeltpunt: –129 °C
Dichtheid: 0,852
Vlampunt: 22 °C (71 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002920
CAS nummer: 107-18-6
EINECS: 203-470-7
UN: 1098
ADR: 6.1,I
Merck Index: 14,286

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR