4-(Indol-3-yl)butyric acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 3-Indolebutyric acid, Indole-3-butyric acid

A15260.14 A15260.06 A15260.22
A15260.14EA 173 EUR
A15260.14 A15260.06 A15260.22
4-(Indol-3-yl)butyric acid ≥98%
4-(Indol-3-yl)butyric acid

Purity: 98% CAS No.: 133-32-4 Molecular Formula: C12H13NO2 Formula Weight: 203.24 Odour: Odourless Melting Point: 121-125° Beilstein Reference: 171120 Merck Reference: 14,4965 UN No.: 2811 MDL No.: MFCD00005664

Formule: C₁₂H₁₃NO₂
Smeltpunt: 121…125 °C
Dichtheid: 1,27 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005664
CAS nummer: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,4965

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR