3,5-Dichloorfenol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A15328.06 A15328.14 A15328.18
A15328.06EA 55.8 EUR
A15328.06 A15328.14 A15328.18
3,5-Dichloorfenol ≥98%
3,5-Dichloorfenol

Purity: 98% CAS No.: 591-35-5 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 63-68° Boiling Point: 233° Beilstein Reference: 1906458 UN No.: 2020 MDL No.: MFCD00002259

Formule: C₆H₄Cl₂O
MW: 163 g/mol
Kookpunt: 233 °C
Smeltpunt: 63…68 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002259
CAS nummer: 591-35-5
EINECS: 209-714-9
UN: 2020
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR