Bromotrimethylsilane ≥97% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Stabilisatie: Stabilised with copper

A15334.09 A15334.18 A15334.30
A15334.09EA 41.4 EUR
A15334.09 A15334.18 A15334.30
Bromotrimethylsilane ≥97% stabilised
Bromotrimethylsilane

Stabilized with copper powder or silver wire


Purity: 97% CAS No.: 2857-97-8 Molecular Formula: C3H9BrSi Formula Weight: 153.09 Storage Temperature: 2°C to 8°C Flash Point: -3°(26°F) Refractive Index: 1.4250 Odour: Acrid Melting Point: -44° Boiling Point: 78-80° Beilstein Reference: 1731135 UN No.: 2924 MDL No.: MFCD00000048

Formule: C₃H₉BrSi
Kookpunt: 78…80 °C
Smeltpunt: –44 °C
Dichtheid: 1,183
Vlampunt: –3 °C (26 °F)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000048
CAS nummer: 2857-97-8
EINECS: 220-672-0
UN: 2924
ADR: 3,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR