1-Chloor-2-nitrobenzeen ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1-Chloro-2-nitrobenzene, 2-Chloronitrobenzene

A15403.0B A15403.0I A15403.30
A15403.0BEA 61.2 EUR
A15403.0B A15403.0I A15403.30
1-Chloor-2-nitrobenzeen ≥99%
1-Chloor-2-nitrobenzeen

Purity: 99% CAS No.: 88-73-3 Molecular Formula: C6H4ClNO2 Formula Weight: 157.56 Flash Point: 126°(258°F) Melting Point: 31-34° Boiling Point: 245-246° Beilstein Reference: 509273 Merck Reference: 14,2151 UN No.: 1578 MDL No.: MFCD00007061

Formule: C₆H₄ClNO₂
MW: 157.56 g/mol
Kookpunt: 245…246 °C
Smeltpunt: 31…34 °C
Dichtheid: 1.348
Vlampunt: 126 °C (258 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007061
CAS nummer: 88-73-3
EINECS: 201-854-9
UN: 1578
ADR: 6.1 II
Merck Index: 13,02170

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR