Picolinamide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A15410.06 A15410.14
A15410.06EA 49.5 EUR
A15410.06 A15410.14
Picolinamide ≥98%
Picolinamide

Purity: 98% CAS No.: 1452-77-3 Molecular Formula: C6H6N2O Formula Weight: 122.13 Melting Point: 104-108° Boiling Point: 143°/20mm MDL No.: MFCD00023483

Formule: C₆H₆N₂O
Kookpunt: 143 °C (20 mmHg)
Smeltpunt: 104…108 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00023483
CAS nummer: 1452-77-3
EINECS: 215-921-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR