Benzotriazole ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A15423.18 A15423.0B A15423.30
A15423.18EA 31.9 EUR
A15423.18 A15423.0B A15423.30
Benzotriazole ≥99%
Benzotriazole

Purity: 99% CAS No.: 95-14-7 Molecular Formula: C6H5N3 Formula Weight: 119.13 Flash Point: 212°(413°F) Physical Form: Powder Density: 1.360 Appearance: White to pale cream Odour: Odourless pH: 6.0-7.0 Melting Point: 96-99° Boiling Point: 204°/15mm Beilstein Reference: 112133 Merck Reference: 14,1108 MDL No.: MFCD00005699

Formule: C₆H₅N₃
Kookpunt: 204 °C (15 mmHg)
Smeltpunt: 96…99 °C
Dichtheid: 1,360
Vlampunt: 212 °C (413 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005699
CAS nummer: 95-14-7
EINECS: 202-394-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,1108
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba54cc42ef5ad9dbae6d7f118a3af1ae642287&vid=ce57b3f0d2583e32220d1e191d0202ad452e6475&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR