2-Nitrobenzyl alcohol ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A15496.14 A15496.22 A15496.36
A15496.14EA 49.8 EUR
A15496.14 A15496.22 A15496.36
2-Nitrobenzyl alcohol ≥97%
2-Nitrobenzyl alcohol

Purity: 97% CAS No.: 612-25-9 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 70-74° Boiling Point: 270° MDL No.: MFCD00007186

Formule: C₇H₇NO₃
Kookpunt: 270 °C
Smeltpunt: 70…74 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007186
CAS nummer: 612-25-9
EINECS: 210-302-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR