Phthalide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A15516.22 A15516.36
A15516.22EA 33.2 EUR
A15516.22 A15516.36
Phthalide ≥98%
Phthalide

Purity: 98+% CAS No.: 87-41-2 Molecular Formula: C8H6O2 Formula Weight: 134.13 Flash Point: 152°(305°F) Melting Point: 72-75° Boiling Point: 289-291° Beilstein Reference: 114632 MDL No.: MFCD00005906

Formule: C₈H₆O₂
Kookpunt: 289…291 °C
Smeltpunt: 72…75 °C
Dichtheid: 1,1
Vlampunt: 152 °C (305 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005906
CAS nummer: 87-41-2
EINECS: 201-744-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR