2-Thiophenesulphonyl chloride ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A15553.06 A15553.14
A15553.06EA 62.7 EUR
A15553.06 A15553.14
2-Thiophenesulphonyl chloride ≥97%
2-Thiophenesulphonyl chloride

Purity: 97% CAS No.: 16629-19-9 Molecular Formula: C4H3ClO2S2 Formula Weight: 182.65 Flash Point: 110°(230°F) Refractive Index: 1.5730 Melting Point: 29-31° Boiling Point: 80-81°/1mm Beilstein Reference: 123536 UN No.: 3261 MDL No.: MFCD00005426

Formule: C₄H₃ClO₂S₂
Kookpunt: 80…81 °C (1 mmHg)
Smeltpunt: 29…31 °C
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005426
CAS nummer: 16629-19-9
EINECS: 240-677-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR