Allantoin ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A15571.30 A15571.0B A15571.0I
A15571.30EA 51.3 EUR
A15571.30 A15571.0B A15571.0I
Allantoin ≥98%
Allantoin

Purity: 98% CAS No.: 97-59-6 Molecular Formula: C4H6N4O3 Formula Weight: 158.12 Melting Point: ca 230° dec. Beilstein Reference: 102364 Merck Reference: 14,258 MDL No.: MFCD00005260

Formule: C₄H₆N₄O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005260
CAS nummer: 97-59-6
EINECS: 202-592-8
Merck Index: 14,258

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR