3-Phenoxybenzyl chloride ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A15670.03 A15670.06 A15670.14
A15670.03EA 42 EUR
A15670.03 A15670.06 A15670.14
3-Phenoxybenzyl chloride ≥97%
3-Phenoxybenzyl chloride

Purity: 97% CAS No.: 53874-66-1 Molecular Formula: C13H11ClO Formula Weight: 218.69 Flash Point: 110°(230°F) Refractive Index: 1.5910 Boiling Point: 141-142°/5mm Beilstein Reference: 2212101 UN No.: 3265 MDL No.: MFCD00040866

Formule: C₁₃H₁₁ClO
Kookpunt: 141…142 °C (5 mmHg)
Dichtheid: 1,189
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00040866
CAS nummer: 53874-66-1
EINECS: 258-831-1
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR