(±)-Epichloorhydrine ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A15823.36 A15823.0E A15823.22
A15823.36EA 34 EUR
A15823.36 A15823.0E A15823.22
(±)-Epichloorhydrine ≥99%
(±)-Epichloorhydrine

Purity: 99% CAS No.: 106-89-8 Molecular Formula: C3H5ClO Formula Weight: 92.53 Flash Point: 32°(90°F) Physical Form: Liquid Density: 1.182 Refractive Index: 1.4380 Appearance: Colourless Odour: Like chlorine Melting Point: -57° Boiling Point: 115-117° Beilstein Reference: 79785 Merck Reference: 14,3611 UN No.: 2023 MDL No.: MFCD00005132


A cyclodextrin cross-linking reagent

Formule: C₃H₅ClO
Kookpunt: 115…117 °C
Smeltpunt: –57 °C
Dichtheid: 1,182
Vlampunt: 32 °C (90 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005132
CAS nummer: 106-89-8
EINECS: 203-439-8
UN: 2023
ADR: 6.1,II
Merck Index: 14,3611

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR