4-Bromo-2-fluorotoluene ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A15891.06 A15891.14 A15891.22
A15891.06EA 26.9 EUR
A15891.06 A15891.14 A15891.22
4-Bromo-2-fluorotoluene ≥99%
4-Bromo-2-fluorotoluene

Purity: 99% CAS No.: 51436-99-8 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 73°(163°F) Refractive Index: 1.5290 Boiling Point: 66-68°/8mm Beilstein Reference: 1859028 MDL No.: MFCD00013551

Formule: C₇H₆BrF
Kookpunt: 66…68 °C (8 mmHg)
Dichtheid: 1,492
Vlampunt: 73 °C (163 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013551
CAS nummer: 51436-99-8
EINECS: 257-201-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR