L-Valine methyl ester hydrochloride 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A15892.09 A15892.18
A15892.09EA 60.6 EUR
A15892.09 A15892.18
L-Valine methyl ester hydrochloride 99%
L-Valine methyl ester hydrochloride

Purity: 99% CAS No.: 6306-52-1 Molecular Formula: C6H13NO2.HCl Formula Weight: 167.64 Storage Temperature: 2°C to 8°C Melting Point: 166° dec. Beilstein Reference: 3594960 Optical Rotation: +16° (c=2 in water) MDL No.: MFCD00012497

Formule: C₆H₁₄ClNO₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012497
CAS nummer: 6306-52-1
EINECS: 228-620-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR