Thiophenol ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A15916.0B A15916.18 A15916.30 A15916.99
A15916.0BEA 157 EUR
A15916.0B A15916.18 A15916.30 A15916.99
Thiophenol ≥99%
Thiophenol

Purity: 99+% CAS No.: 108-98-5 Molecular Formula: C6H6S Formula Weight: 110.18 Flash Point: 55°(131°F) Physical Form: Liquid Density: 1.075 Refractive Index: 1.5900 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Odour: Stench pH: 5 Melting Point: -15° Boiling Point: 168-169° Beilstein Reference: 506523 Merck Reference: 14,9355 UN No.: 2337 MDL No.: MFCD00004826

Formule: C₆H₆S
Kookpunt: 168…169 °C
Smeltpunt: –15 °C
Dichtheid: 1,075
Vlampunt: 55 °C (131 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004826
CAS nummer: 108-98-5
EINECS: 203-635-3
UN: 2337
ADR: 6.1,I
Merck Index: 14,9355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR